Oxalic acid 0.005 mol/l (0.01 N), AVS TITRINORM Volumetric

Supplier: VWR Chemicals

Synonyms: Ethanedioic acid

89619.290EA 49.2 EUR
89619.290
Oxalic acid 0.005 mol/l (0.01 N), AVS TITRINORM Volumetric
Oxalic acid
Formula: C₂H₂O₄
MW: 90.04 g/mol
Boiling Pt: 365
MDL Number: MFCD00002573
CAS Number: 144-62-7
Merck Index: 13,06980

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR