Potassium bis(oxalato)oxotitanate(IV) dihydrate

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Dipotassium oxodioxalatotitanate(IV) dihydrate, Potassium titanium oxide oxalate dihydrate

89620.36 89620.A3 89620.PG
89620.36EA 129 EUR
89620.36 89620.A3 89620.PG
Potassium bis(oxalato)oxotitanate(IV) dihydrate
Potassium bis(oxalato)oxotitanate(IV) dihydrate

CAS No.: 14402-67-6 Molecular Formula: K2[TiO(C2O4)2].2H2O Formula Weight: 354.10 (318.10anhy) Physical Form: Crystalline Appearance: Colourless Odour: Odourless Solubility: Soluble in water UN No.: UN3288 MDL No.: MFCD00150592


Applications: Mordant in dyeing, reagent

Formula: K₂[TiO(C₂O₄)₂]·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150592
CAS Number: 14402-67-6
EINECS: 238-475-3
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR