Lithium aluminum hydride ≥97%, pellets

Supplier: Thermo Fisher Scientific
Danger

89643.14 89643.36
89643.14EA 73.2 EUR
89643.14 89643.36
Lithium aluminum hydride ≥97%, pellets
Lithium aluminum hydride

Purity: 97% CAS No.: 16853-85-3 Molecular Formula: LiAlH4 Formula Weight: 37.95 Physical Form: Pellets Density: 0.917 Appearance: Whote to grey Odour: Odourless Melting Point: 125° dec. Merck Reference: 14,349 UN No.: 1410 MDL No.: MFCD00011075

Formula: H₄AlLi
Melting Pt: >125 °C (dec.)
Density: 0,917
Storage Temperature: Ambient
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1410
ADR: 4.3,I
Merck Index: 14,349

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR