VWR® Hitachi Empower 3 Chromatography Data Software
Supplier: VWR Collection
906-0156EA 2710 EUR
906-0156 HITA892-0011S HITA892-0012S
VWR® Hitachi Empower 3 Chromatography Data Software
Software Chromatography Software
Empower 3 is Waters's compliant-ready chromatography data software (CDS) package for advanced data acquisition, management, processing, reporting and distribution. Empower 3's interface is designed to maximise laboratory efficiency, improving collection, processing and printing chromatography data. Capabilities include:
  • Customisable data reports
  • Integrated custom calculations
  • Relational data base so all the meta data is traceable back to the raw data
  • Optimised workflow for different users and different analyses
With enhanced 21 CFR Part 11 complianceMore Product Information