Supelco® Organic Reference Standard, Eleutheroside B

Supplier: Merck

90974-10MG
90974-10MGEA 531 EUR
90974-10MG
Supelco® Organic Reference Standard, Eleutheroside B
Standards Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edce0d86f3f31c0b31765d44a786fdb42959205e&vid=c4e3083a92c1a9396e1ff70d1d9adb9d4551d2ba&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) (E)-4-[-3-Hydroxy-1-propenyl]-2,6-dimethoxyphenyl-β-ᴅ-glucopyranoside; Ilexanthin A; Ligustrin; Lilacin; Magnolenin; Methoxyconiferine; Sinapyl alcohol 4-O-glucoside; Syringin; Syringoside
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 118-34-3
MDL No. MFCD00016778
Molecular weight 372,37
Shipping conditions STD
Storage conditions 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR