Methyl tridecanoate, Supelco®

Supplier: Merck

Synonyms: Tridecylic acid methyl ester, Tridecanoic acid methyl ester

M-Clarity™ quality level = MQ100

91558-5ML
91558-5MLEA 255 EUR
91558-5ML
Methyl tridecanoate, Supelco®
Methyl tridecanoate
Formula: C₁₄H₂₈O₂
MW: 228.38 g/mol
Boiling Pt: 290
Melting Pt: 5.5
Density: 0.864 g/cm³ (25 °C)
MDL Number: MFCD00008977
CAS Number: 1731-88-0
EINECS: 217-054-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=324c85267054efe393f254586ab27221a8e878e3&vid=0536e12a64753d922a72d42f8d10c9fb51d57a19&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR