Microplates, 1536-well
Supplier: HIRSCHMANN
9240136
9240136.EA 0 EUR
9240136.
Microplates, 1536-well
Microplates Microplates with Glass Inserts
Microtitration plate, made from PP, with fixed glass inserts.
  • High purity and chemically inert
  • 1536 wells
  • Translucent plastic cover included in standard scope of delivery
Plate dimensions: 124×81 mm