Pasteur pipettes
Supplier: HIRSCHMANN
9250101 9250201 9260101 9260201
9250101.EA 66.4 EUR
9250101. 9250201. 9260101. 9260201.
Pasteur pipettes
Pipettes Pasteur/Transfer Pipettes
Glass pasteur pipettes with fine drawn tips.
  • With or without cotton plug
  • Short or long form
Certifications: According to ISO 7712.
Order Now
SPECIFICATIONS
Environmentally Preferable