TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Supelco®

Supplier: Merck

Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

M-Clarity™ quality level = MQ100

93313-1L
93313-1LEA 142 EUR
93313-1L
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Supelco®
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)
Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28ff15da07c66645899ad400511069f72a82cbbd&vid=5f71031df0f00dfd45ae10782505e7221edcea4b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR