Spatulas, Trulla type
Supplier: NICKEL ELECTRO
9370
9370.EA 54.4 EUR
9370.
Spatulas, Trulla type
Spatulas
Stainless steel 18/8 non-magnetic, bright finish. Double ended spatulas with trowel shaped scoops for handling large quantities of material.
  • Excellent thermal resistance, autoclavable
  • Excellent chemical resistance
  • Quick and easy to clean