Supelco® TraceCERT® Organic Reference Standard, Bromopropylate

Supplier: Merck

93744-50MG
93744-50MGEA 69.5 EUR
93744-50MG
Supelco® TraceCERT® Organic Reference Standard, Bromopropylate
Standards Organic Standards

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Bromopropylate CRM may also find following uses:


  • To elucidate the inhibition of plasma and brain acetylcholinesterase activity in Wistar rats with a mixture of 5 pesticides: chlorpyrifos, alphacypermetrin, bromopropylate, carbendazim, and mancozeb
  • Study the degradation processes of bromopropylate, coumaphos, chlordimeform, cymiazole, flumethrin, and fluvalinate in aqueous media by HPLC
  • To develop a liquid-phase microextraction method using deep eutectic solvent for extraction and preconcentration of diazinon, metalaxyl, bromopropylate, oxadiazon, and fenazaquin pesticides from samples such as grape and sour cherry juices, fresh beet, cucumber, potato, and tomato followed by gas chromatography-flame ionisation detection
  • To develop a multi-residue method for the determination of azinphos methyl, bromopropylate, chlorpyrifos, dimethoate, parathion methyl, and phosalone in apricots and peaches using negative chemical ionisation ion trap technology
  • Simultaneous determination of amitraz, bromopropylate, coumaphos, cymiazole, and fluvalinate residues in honey by solid-phase extraction and GC-MS


Caution: P273 - P391 - P501

Order Now

SPECIFICATIONS

Synonym(s) Isopropyl 4,4′-dibromobenzilate; Phenisobromolate
Environmentally Preferable
M-Clarity quality level MQ300
CAS No. 18181-80-1
MDL No. MFCD00078632
Molecular weight 428,12
Hazard class description H410
Storage conditions 15...25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR