o-Xylene, Supelco®

Supplier: Merck
Danger

Synonyms: 1,2-Dimethylbenzene

M-Clarity™ quality level = MQ100

95660-5ML 95660-50ML 95660-5ML
95660-5MLEA 105 EUR
95660-5ML 95660-50ML NOVS95660-5ML
o-Xylene, Supelco®
o-Xylene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 144
Melting Pt: –25 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 30 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008519
CAS Number: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3 III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=950bdbbeadd692c67ac79579692ca8939279051e&vid=b5b5e2f450177427f036dbac13f0ae470d2dcbd3&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR