Avantor® Seradigm, Foetal Bovine Serum (FBS), US origin
Supplier: VWR Seradigm
89510-196EA 121 EUR
89510-196 97068-102 97068-107 97068-086 89510-194 97068-085 89510-198 97068-101 89510-200 97068-088 97068-091 97068-104
Avantor® Seradigm, Foetal Bovine Serum (FBS), US origin
Sera (Serum)
Ultimate grade FBS offers the ultimate in product quality and viral safety, undergoing additional BVD virus testing using qPCR technology. Premium grade FBS is widely used by cell culturists requiring high quality, high performance FBS for a wide variety of applications.
  • 100% US origin
  • Triple 0,1 μm sterile filtration
  • Preservation of native, growth-promoting factors and minimisation of growth-inhibiting factors
  • Independently tested by third party laboratories
  • Fully traceable
Ultimate grade FBS (endotoxin ≤5 EU/ml; haemogloblin ≤25 mg/dl) offers theMore Product Information