VWR®, Cone and Plate viscosity standards
Supplier: VWR Chemicals
99406.260EA 201 EUR
99406.260 99407.260 99408.260 99409.260 99411.260 99412.260 99413.260 99414.260 99415.260 99417.260 99418.260 99419.260 99421.260 99422.260 99423.260 99424.260 99425.260 99426.260 99427.260
VWR®, Cone and Plate viscosity standards
Standards Viscometry Standards
High quality Cone and Plate viscosity standards are ideal for the paints and coatings industry.
  • Kinematic viscosity certified in strict accordance with ASTM D2162 at 20, 23 and 25 °C
  • Kinematic viscosity certified in accordance with ASTM D341 at intermediate temperatures
  • Density, dynamic viscosity and kinematic viscosity quoted at all temperatures
  • Ensures full compliance to ASTM and IP test method protocols and had full traceability to national standards
  • Tamper-evident security packaging
Certifications: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
Caution: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.