Pipette tray
Supplier: KARTELL
996
996.EA 57.8 EUR
996.
Pipette tray
Trays/Dishes/Pots Pipette Trays
PVC.
  • Very useful workstation holder for items other than pipettes
  • For 7 pipettes diameter <20 mm or for 16 pipettes diameter <10 mm
  • For pipettes of all lengths
W×D×H: 283×216×40 mm