Cadmium(II) sulphate hydrate 98% (by complexometric analysis)

Supplier: Thermo Fisher Scientific
Danger

A10168.0E A10168.22 A10168.36
A10168.0EEA 577 EUR
A10168.0E A10168.22 A10168.36
Cadmium(II) sulphate hydrate 98% (by complexometric analysis)
Cadmium (II) sulphate hydrate

Purity: 98% CAS No.: 15244-35-6 Molecular Formula: 3CdSO4.xH2O (x≈1) Formula Weight: 208.46(anhy) Refractive Index: 1.565 Odour: Odourless Merck Reference: 14,1627 UN No.: 2570 MDL No.: MFCD00151655

Formula: CdO₄S·H₂O
Melting Pt: 41,5 °C trans pt
Density: 3,08
Storage Temperature: Ambient
MDL Number: MFCD00151655
CAS Number: 15244-35-6
EINECS: 233-331-6
UN: 2570
ADR: 6.1,III
Merck Index: 14,1627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR