Sodium perborate tetrahydrate 97

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Sodium peroxometaborate tetrahydrate

A10189.0E A10189.22 A10189.36
A10189.0EEA 78.8 EUR
A10189.0E A10189.22 A10189.36
Sodium perborate tetrahydrate 97
Sodium perborate tetrahydrate

Purity: 97% CAS No.: 10486-00-7 Molecular Formula: NaBO3.4H2O Formula Weight: 153.86 (81.84anhy) Odour: Odourless pH: ca 10 Melting Point: 60° dec. Merck Reference: 14,8652 UN No.: 1479 MDL No.: MFCD00149231

Formula: NaH₂BO₄·4H₂O
MW: 171.87 g/mol
Melting Pt: 60 °C (dec.)
Density: 1.73
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
EINECS: 239-172-9
ADR: 5.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR