2-Amino-3-methylpyridine 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-Methyl-2-pyridylamine, 3-Methylpyridin-2-amine, 2-Amino-3-picoline

A10224.14 A10224.22 A10224.36
A10224.14EA 29.2 EUR
A10224.14 A10224.22 A10224.36
2-Amino-3-methylpyridine 97%
2-Amino-3-methylpyridine

Purity: 97% CAS No.: 1603-40-3 Molecular Formula: C6H8N2 Formula Weight: 108.14 Flash Point: 118°(244°F) Refractive Index: 1.5825 Odour: Unpleasant Melting Point: 29-32° Boiling Point: 221-222° Beilstein Reference: 107892 UN No.: 2811 MDL No.: MFCD00006320

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 221…222 °C
Melting Pt: 29…32 °C
Density: 1.073
Flash Pt: 118 °C (244 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006320
CAS Number: 1603-40-3
EINECS: 216-501-4
UN: 2811
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR