Dibromomethane ≥99% stabilised

Supplier: Thermo Fisher Scientific
Warning


Stabilisation: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A10456.0E A10456.22 A10456.36
A10456.0EEA 202 EUR
A10456.0E A10456.22 A10456.36
Dibromomethane ≥99% stabilised
Dibromomethane

Stabilized with 50 ppm BHT


Purity: 99% CAS No.: 74-95-3 Molecular Formula: CH2Br2 Formula Weight: 173.85 Refractive Index: 1.5410 Melting Point: -53° Boiling Point: 96-98° Beilstein Reference: 969143 Merck Reference: 14,6061 UN No.: 2664 MDL No.: MFCD00000168

Formula: Br₂CH₂
Boiling Pt: 96…98 °C
Melting Pt: –53 °C
Density: 2,495
Storage Temperature: Ambient
MDL Number: MFCD00000168
CAS Number: 74-95-3
EINECS: 200-824-2
UN: 2664
ADR: 6.1,III
Merck Index: 14,6061

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR