Lithium chloride, anhydrous 98+%

Supplier: Thermo Fisher Scientific
Warning

A10531.0E A10531.22 A10531.36
A10531.0EEA 673 EUR
A10531.0E A10531.22 A10531.36
Lithium chloride, anhydrous 98+%
Lithium chloride

Purity: 98+% CAS No.: 7447-41-8 Molecular Formula: LiCl Formula Weight: 42.39 Odour: Odourless Melting Point: 605° Boiling Point: 1325-1360° Merck Reference: 14,5528 MDL No.: MFCD00011078

Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2.068
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR