Methyl-4-(bromomethyl)benzoate ≥98%

Supplier: Thermo Fisher Scientific
Danger

A10557.06 A10557.14 A10557.22
A10557.06EA 71.1 EUR
A10557.06 A10557.14 A10557.22
Methyl-4-(bromomethyl)benzoate ≥98%
Methyl 4-(bromomethyl)benzoate

Purity: 98% CAS No.: 2417-72-3 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Melting Point: 53-58° Boiling Point: 130-135°/2mm UN No.: 1759 MDL No.: MFCD00032453

Formula: C₉H₉BrO₂
Boiling Pt: 130…135 °C (2 mmHg)
Melting Pt: 53…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00032453
CAS Number: 2417-72-3
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR