Ethyl decanoate 99

Supplier: Thermo Fisher Scientific

Synonyms: Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester

A10657.0E A10657.22 A10657.36
A10657.0EEA 332 EUR
A10657.0E A10657.22 A10657.36
Ethyl decanoate 99
Ethyl decanoate

Purity: 99% CAS No.: 110-38-3 Molecular Formula: CH3(CH2)8CO2CH2CH3 Formula Weight: 200.32 Flash Point: 102°(216°F) Refractive Index: 1.4250 Melting Point: -20° Boiling Point: 250-252° Beilstein Reference: 1762128 Merck Reference: 14,3776 MDL No.: MFCD00009581

Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 250…252 °C
Melting Pt: –20 °C
Density: 0.862
Flash Pt: 102 °C (216 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009581
CAS Number: 110-38-3
EINECS: 203-761-9
Merck Index: 13,03811

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR