α-Bromopropiophenone 96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Bromopropiophenone

A10661.14 A10661.22 A10661.36
A10661.14EA 62 EUR
A10661.14 A10661.22 A10661.36
α-Bromopropiophenone 96%
α-Bromopropiophenone

Purity: 96% CAS No.: 2114-00-3 Molecular Formula: C9H9BrO Formula Weight: 213.08 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5720 Odour: Pungent Boiling Point: 245-250° Beilstein Reference: 508550 UN No.: 2927 MDL No.: MFCD00000145

Formula: C₆H₅C(O)CHBrCH₃
Boiling Pt: 245…250 °C
Density: 1,417
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000145
CAS Number: 2114-00-3
EINECS: 218-307-5
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR