EDTA (ethylenediamine tetraacetic acid) 99%

Supplier: Thermo Fisher Scientific
Warning

A10713.0B A10713.0I A10713.30 A10713.36
A10713.0BEA 79 EUR
A10713.0B A10713.0I A10713.30 A10713.36
EDTA (ethylenediamine tetraacetic acid) 99%
EDTA (Ethylenediamine tetraacetic acid)

Purity: 99% CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Melting Point: ca 245° dec. Beilstein Reference: 1716295 Merck Reference: 14,3517 MDL No.: MFCD00003541

Formula: C₁₀H₁₆N₂O₈
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: ∼ 245 °C (dec.)
Density: 0,86 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 14,3517
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR