2-Chloro-α,α,α-trifluorotoluene ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Chloro-2-(trifluoromethyl)benzene, 2-Chlorobenzotrifluoride

A10753.0E A10753.22 A10753.36
A10753.0EEA 279 EUR
A10753.0E A10753.22 A10753.36
2-Chloro-α,α,α-trifluorotoluene ≥99%
2-Chloro-α,α,α-trifluorotoluene

Purity: 99% CAS No.: 88-16-4 Molecular Formula: C7H4ClF3 Formula Weight: 180.56 Flash Point: 58°(136°F) Refractive Index: 1.4560 Melting Point: -6° Boiling Point: 151-153° Beilstein Reference: 510993 UN No.: 2234 MDL No.: MFCD00000561

Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Boiling Pt: 151…153 °C
Melting Pt: –6 °C
Density: 1.369
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000561
CAS Number: 88-16-4
EINECS: 201-805-1
UN: 2234
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR