α-Chloro-m-xylene ≥98%

Supplier: Thermo Fisher Scientific
Danger

A10784.14 A10784.22 A10784.36
A10784.14EA 34.7 EUR
A10784.14 A10784.22 A10784.36
α-Chloro-m-xylene ≥98%
α-Chloro-m-xylene

Purity: 98% CAS No.: 620-19-9 Molecular Formula: C8H9Cl Formula Weight: 140.61 Flash Point: 75°(167°F) Refractive Index: 1.5350 Odour: Pungent Boiling Point: 195-196° Beilstein Reference: 2040357 Merck Reference: 14,10091 UN No.: 3265 MDL No.: MFCD00000909

Formula: C₈H₉Cl
Boiling Pt: 195…196 °C
Density: 1,066
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000909
CAS Number: 620-19-9
EINECS: 210-628-9
UN: 3265
ADR: 8,II
Merck Index: 14,10091

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR