2-Phenoxyethanol ≥94%

Supplier: Thermo Fisher Scientific
Warning

A10786.0B A10786.0I A10786.30
A10786.0BEA 68.8 EUR
A10786.0B A10786.0I A10786.30
2-Phenoxyethanol ≥94%
2-Phenoxyethanol
Formula: C₆H₅OCH₂CH₂OH
Boiling Pt: 245 °C
Melting Pt: 11…14 °C
Density: 1,102
Flash Pt: 121 °C (249 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002857
CAS Number: 122-99-6
EINECS: 204-589-7
Merck Index: 14,7257
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27cc4d39b806528798164961c23a1285d41d6a9&vid=a46b721cc69101d0c50dc49d9687445b395a3766&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR