Tricine

Supplier: PanReac AppliChem

A1085.0250 A1085.1000 A1085.5000
A1085.0250EA 238 EUR
A1085.0250 A1085.1000 A1085.5000
Tricine
Tricine
Formula: (HOCH₂)₃CNHCH₂CO₂H
Melting Pt: 187 °C
Density: 1,523 g/cm³ (19.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR