2,3,5-Triphenyltetrazolium chloride (TTC) ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC

A10870.09 A10870.18 A10870.30
A10870.09EA 68.4 EUR
A10870.09 A10870.18 A10870.30
2,3,5-Triphenyltetrazolium chloride (TTC) ≥98%
2,3,5-Triphenyltetrazolium chloride

Purity: 98% CAS No.: 298-96-4 Molecular Formula: C19H15ClN4 Formula Weight: 334.81 Storage Temperature: 2°C to 8°C Physical Form: Crystals Sensitivity: Light Sensitive and Hygroscopic Appearance: White to pale yellow Melting Point: ca 250° dec. Beilstein Reference: 3923356 Merck Reference: 14,9744 MDL No.: MFCD00011963


Useful indicator for reducing substances

Formula: C₁₉H₁₅ClN₄
MW: 334.81 g/mol
Boiling Pt: ∼360 °C (1013 hPa)
Melting Pt: ∼ 250 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011963
CAS Number: 298-96-4
EINECS: 206-071-6
Merck Index: 14,9744

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR