Aluminium potassium sulphate dodecahydrate 98

Supplier: Thermo Fisher Scientific

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

A10906.0E A10906.36
A10906.0EEA 109 EUR
A10906.0E A10906.36
Aluminium potassium sulphate dodecahydrate 98
Aluminium potassium sulphate dodecahydrate

Purity: 98+% CAS No.: 7784-24-9 Molecular Formula: KAl(SO4)2.12H2O Formula Weight: 474.38 (258.20anhy) Odour: Odourless Melting Point: 92° MDL No.: MFCD00149143

Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92 °C
Density: 1.757
Storage Temperature: Ambient
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR