2,4-Dimethyl-6-hydroxypyrimidine 99%

Supplier: Thermo Fisher Scientific
Warning

A10960.06 A10960.14
A10960.06EA 30.6 EUR
A10960.06 A10960.14
2,4-Dimethyl-6-hydroxypyrimidine 99%
2,4-Dimethyl-6-hydroxypyrimidine

Purity: 99% CAS No.: 6622-92-0 Molecular Formula: C6H8N2O Formula Weight: 124.14 Melting Point: 198-200° Beilstein Reference: 114324 MDL No.: MFCD00006105

Formula: C₆H₈N₂O
Melting Pt: 198…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00006105
CAS Number: 6622-92-0
EINECS: 229-577-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR