4-Chlorothioanisole ≥98%

Supplier: Thermo Fisher Scientific
Warning

A11025.06 A11025.14
A11025.06EA 42.6 EUR
A11025.06 A11025.14
4-Chlorothioanisole ≥98%
4-Chlorothioanisole

Purity: 98% CAS No.: 123-09-1 Molecular Formula: C7H7ClS Formula Weight: 158.65 Flash Point: 90°(194°F) Refractive Index: 1.600 Odour: Unpleasant Boiling Point: 223° Beilstein Reference: 2041509 MDL No.: MFCD00013643

Formula: C₇H₇ClS
Boiling Pt: 223 °C
Melting Pt: 27…28 °C
Density: 1,220
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013643
CAS Number: 123-09-1
EINECS: 204-600-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR