Diisoamyl ether (mixture of isomers) 96%

Supplier: Thermo Fisher Scientific
Warning

A11231.AC A11231.AE A11231.AP
A11231.ACEA 30.4 EUR
A11231.AC A11231.AE A11231.AP
Diisoamyl ether (mixture of isomers) 96%
Diisoamyl ether (mixture of isomers)

Purity: 96% CAS No.: 544-01-4 Molecular Formula: [(CH3)2CHCH2CH2]2O Formula Weight: 158.29 Flash Point: 46°(115°F) Refractive Index: 1.407 Boiling Point: 172-174° Beilstein Reference: 1698014 Merck Reference: 14,5118 UN No.: 3271 MDL No.: MFCD00008947

Formula: C₁₀H₂₂O
MW: 158.28 g/mol
Boiling Pt: 172…174 °C
Density: 0.78
Flash Pt: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008947
CAS Number: 544-01-4
EINECS: 208-857-4
UN: 3271
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR