trans-2,3-Dimethoxy-β-nitrostyrene ≥98%

Supplier: Thermo Fisher Scientific
Warning

A11347.06
A11347.06EA 0 EUR
A11347.06
trans-2,3-Dimethoxy-β-nitrostyrene ≥98%
trans-2,3-Dimethoxy-β-nitrostyrene

Purity: 98% CAS No.: 37630-20-9 Molecular Formula: C10H11NO4 Formula Weight: 209.19 Melting Point: 80-82° MDL No.: MFCD00068082

Formula: C₁₀H₁₁NO₄
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00068082
CAS Number: 37630-20-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR