Phenol equilibrated 89 - 90% 89-90 Stabilised

Supplier: PanReac AppliChem
Danger


Stabilisation: 8-Hydroxychinolin

A1153.0500
A1153.0500EA 226 EUR
A1153.0500
Phenol equilibrated 89 - 90% 89-90 Stabilised
Phenol equilibrated
Formula: C₆H₅OH
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 2821
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR