α,α'-Dichloro-m-xylene ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1,3-Bis(chloromethyl)benzene, m-Xylene dichloride, m-Xylylene dichloride

A11548.09 A11548.14 A11548.22
A11548.09EA 50.9 EUR
A11548.09 A11548.14 A11548.22
α,α'-Dichloro-m-xylene ≥97%
α,α'-Dichloro-m-xylene

Purity: 97% CAS No.: 626-16-4 Molecular Formula: C8H8Cl2 Formula Weight: 175.06 Flash Point: 144°(291°F) Melting Point: 33-37° Boiling Point: 250-255° Beilstein Reference: 2079898 UN No.: 2928 MDL No.: MFCD00000912

Formula: C₈H₈Cl₂
MW: 175.06 g/mol
Boiling Pt: 250…255 °C
Melting Pt: 33…37 °C
Density: 1.302
Flash Pt: 144 °C (291 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000912
CAS Number: 626-16-4
EINECS: 210-932-1
UN: 2928
ADR: 6.1 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR