2,4-Difluorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Danger

A11667.06 A11667.14
A11667.06EA 21.6 EUR
A11667.06 A11667.14
2,4-Difluorotoluene ≥98%
2,4-Difluorotoluene

Purity: 98+% CAS No.: 452-76-6 Molecular Formula: C7H6F2 Formula Weight: 128.12 Flash Point: 15°(59°F) Refractive Index: 1.4490 Odour: Aromatic Boiling Point: 114-115° Beilstein Reference: 1931681 UN No.: 2388 MDL No.: MFCD00000332

Formula: C₇H₆F₂
Boiling Pt: 114…115 °C
Density: 1,121
Flash Pt: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000332
CAS Number: 452-76-6
EINECS: 207-211-9
UN: 2388
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR