Sanger's reagent ≥99%

Supplier: Thermo Fisher Scientific
Danger

A11871.14 A11871.06 A11871.22
A11871.14EA 95.8 EUR
A11871.14 A11871.06 A11871.22
Sanger's reagent ≥99%
Sanger's reagent

Purity: 99% CAS No.: 70-34-8 Molecular Formula: C6H3FN2O4 Formula Weight: 186.10 Flash Point: 110°(230°F) Refractive Index: 1.5690 Melting Point: 24-30° Boiling Point: 296° Beilstein Reference: 398632 Merck Reference: 14,4172 UN No.: 2811 MDL No.: MFCD00007056

Formula: C₆H₃FN₂O₄
Boiling Pt: 296 °C
Melting Pt: 24…30 °C
Density: 1,482
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007056
CAS Number: 70-34-8
EINECS: 200-734-3
UN: 2811
ADR: 6.1,III
Merck Index: 14,4172

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR