N-Benzoyl-N-phenylhydroxylamine 98%

Supplier: Thermo Fisher Scientific

Synonyms: Tantalon, N-Phenylbenzohydroxamic Acid, N-Hydroxy-N-phenylbenzamide

A11950.06 A11950.14
A11950.06EA 115 EUR
A11950.06 A11950.14
N-Benzoyl-N-phenylhydroxylamine 98%
N-Benzoyl-N-phenylhydroxylamine

Purity: 98% CAS No.: 304-88-1 Molecular Formula: C13H11NO2 Formula Weight: 213.24 Melting Point: 118-120° Beilstein Reference: 2212449 MDL No.: MFCD00002111

Formula: C₁₃H₁₁NO₂
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00002111
CAS Number: 304-88-1
EINECS: 206-158-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR