4'-Hydroxyacetophenone ≥99%

Supplier: Thermo Fisher Scientific
Danger

A11974.0B A11974.22 A11974.36
A11974.0BEA 269 EUR
A11974.0B A11974.22 A11974.36
4'-Hydroxyacetophenone ≥99%
4'-Hydroxyacetophenone

Purity: 99% CAS No.: 99-93-4 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 166°(330°F) Melting Point: 106-108° Boiling Point: 286° Beilstein Reference: 774355 MDL No.: MFCD00002359

Formula: C₈H₈O₂
Boiling Pt: 286 °C
Melting Pt: 106…108 °C
Density: 1,139 g/cm³ (25 °C)
Flash Pt: 166 °C (330 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002359
CAS Number: 99-93-4
EINECS: 202-802-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR