Dichloro(diphenyl)silane ≥97%

Supplier: Thermo Fisher Scientific
Danger

A12051.36 A12051.22
A12051.36EA 173 EUR
A12051.36 A12051.22
Dichloro(diphenyl)silane ≥97%
Dichloro(diphenyl)silane

Purity: 97% CAS No.: 80-10-4 Molecular Formula: C12H10Cl2Si Formula Weight: 253.20 Flash Point: 157°(314°F) Refractive Index: 1.5790 Odour: Pungent Melting Point: -22° Boiling Point: 303-304° Beilstein Reference: 609882 UN No.: 1769 MDL No.: MFCD00000489

Formula: C₁₂H₁₀Cl₂Si
Boiling Pt: 303…304 °C
Melting Pt: –22 °C
Density: 1,208
Flash Pt: 157 °C (314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000489
CAS Number: 80-10-4
EINECS: 201-251-0
UN: 1769
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR