Vanillic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

A12074.14 A12074.22 A12074.36
A12074.14EA 44 EUR
A12074.14 A12074.22 A12074.36
Vanillic acid ≥98%
Vanillic acid

Purity: 98% CAS No.: 121-34-6 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 210-218° Beilstein Reference: 2208364 Merck Reference: 14,9931 MDL No.: MFCD00002551

Formula: C₈H₈O₄
Boiling Pt: 353 °C (1013 hPa)
Melting Pt: 210…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00002551
CAS Number: 121-34-6
EINECS: 204-466-8
Merck Index: 14,9931

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR