Succinic acid ≥99%

Supplier: Thermo Fisher Scientific
Danger

A12084.0C A12084.0E A12084.36
A12084.0CEA 484 EUR
A12084.0C A12084.0E A12084.36
Succinic acid ≥99%
Succinic acid

Purity: 99+% CAS No.: 110-15-6 Molecular Formula: HO2CCH2CH2CO2H Formula Weight: 118.09 Flash Point: 206°(402°F) Melting Point: 186-188° Boiling Point: 235° Beilstein Reference: 1754069 Merck Reference: 14,8869 MDL No.: MFCD00002789

Formula: HOOCCH₂CH₂COOH
Boiling Pt: 235 °C
Melting Pt: 186…188 °C
Density: 1,56
Flash Pt: 206 °C (402 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8,III
Merck Index: 14,8869
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR