Phenylacetylene ≥98%

Supplier: Thermo Fisher Scientific
Danger

A12139.14 A12139.22 A12139.36
A12139.14EA 40.3 EUR
A12139.14 A12139.22 A12139.36
Phenylacetylene ≥98%
Phenylacetylene

Purity: 98+% CAS No.: 536-74-3 Molecular Formula: C8H6 Formula Weight: 102.14 Flash Point: 27°(81°F) Refractive Index: 1.5480 Melting Point: -45° Boiling Point: 142-144° Beilstein Reference: 605461 Merck Reference: 14,3861 UN No.: 3295 MDL No.: MFCD00008570

Formula: C₈H₆
Boiling Pt: 142…144 °C
Melting Pt: –45 °C
Density: 0,930
Flash Pt: 27 °C (81 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008570
CAS Number: 536-74-3
EINECS: 208-645-1
UN: 3295
ADR: 3,III
Merck Index: 14,3861

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR