Salicylic acid ≥99%

Supplier: Thermo Fisher Scientific
Danger

A12253.0E A12253.30 A12253.36
A12253.0EEA 112 EUR
A12253.0E A12253.30 A12253.36
Salicylic acid ≥99%
Salicylic acid

Purity: 99% CAS No.: 69-72-7 Molecular Formula: C7H6O3 Formula Weight: 138.12 Flash Point: 157°(315°F) Melting Point: 158-160° Boiling Point: 211°/20mm Beilstein Reference: 774890 Merck Reference: 14,8332 MDL No.: MFCD00002439

Formula: (HO)C₆H₄CO₂H
Boiling Pt: 211 °C (20 mmHg)
Melting Pt: 158…160 °C
Density: 1,443
Flash Pt: 157 °C (315 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 14,8332
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1f695fa92656a135b5e1742f1cca28f4d255fb&vid=8f3aff6fb9a70e5dd194b8bc46019af74e48e90a&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR