4-Isopropylphenylhydrazine hydrochloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

A12416.03 A12416.06 A12416.14
A12416.03EA 33.2 EUR
A12416.03 A12416.06 A12416.14
4-Isopropylphenylhydrazine hydrochloride ≥98%
4-Isopropylphenylhydrazine hydrochloride

Purity: 98% CAS No.: 118427-29-5 Molecular Formula: C9H14N2.HCl Formula Weight: 186.69 Melting Point: 203° dec. MDL No.: MFCD00013461

Formula: C₉H₁₅ClN₂
Melting Pt: 203 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013461
CAS Number: 118427-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR