3-Fluorobenzyl alcohol ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (3-Fluorophenyl)methanol

A12509.09 A12509.14 A12509.18
A12509.09EA 40 EUR
A12509.09 A12509.14 A12509.18
3-Fluorobenzyl alcohol ≥98%
3-Fluorobenzyl alcohol

Purity: 98% CAS No.: 456-47-3 Molecular Formula: C7H7FO Formula Weight: 126.13 Flash Point: 90°(194°F) Refractive Index: 1.5100 Odour: Characteristic Boiling Point: 201° Beilstein Reference: 2242511 MDL No.: MFCD00004631

Formula: C₇H₇FO
Boiling Pt: 201 °C
Density: 1,166
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004631
CAS Number: 456-47-3
EINECS: 207-265-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR