PTFE

Supplier: Thermo Fisher Scientific

Synonyms: Polytetrafluoroethylene, Teflon

A12613.30 A12613.18 A12613.0B
A12613.30EA 315 EUR
A12613.30 A12613.18 A12613.0B
PTFE
PTFE

CAS No.: 9002-84-0 Molecular Formula: -(CF2CF2-)n MDL No.: MFCD00084455

Formula: (CF₂CF₂)n
Storage Temperature: Ambient
MDL Number: MFCD00084455
CAS Number: 9002-84-0
Merck Index: 14,7562

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR