Trimethylsilyl isocyanate ≥94%

Supplier: Thermo Fisher Scientific
Danger

A12633.09 A12633.18 A12633.30
A12633.09EA 90 EUR
A12633.09 A12633.18 A12633.30
Trimethylsilyl isocyanate ≥94%
Trimethylsilyl isocyanate

Purity: 94% CAS No.: 1118-02-1 Molecular Formula: C4H9NOSi Formula Weight: 115.21 Storage Temperature: -30°C to -10°C Flash Point: -2°(28°F) Refractive Index: 1.3960 Odour: Acrid Melting Point: -49° Boiling Point: 90-92° Beilstein Reference: 1744962 UN No.: 2478 MDL No.: MFCD00001993

Formula: C₄H₉NOSi
Boiling Pt: 90…92 °C
Melting Pt: –49 °C
Density: 0,859
Flash Pt: –2 °C (28 °F)
MDL Number: MFCD00001993
CAS Number: 1118-02-1
EINECS: 214-256-8
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR