Potassium iodide 99

Supplier: Thermo Fisher Scientific

Synonyms: Dipotassium diiodide, Kali iodide

A12704.0B A12704.0I A12704.18 A12704.30
A12704.0BEA 282 EUR
A12704.0B A12704.0I A12704.18 A12704.30
Potassium iodide 99
Potassium iodide

Purity: 99% CAS No.: 7681-11-0 Molecular Formula: KI Formula Weight: 166.01 Refractive Index: 1.677 Odour: Odourless Boiling Point: 1330° Merck Reference: 14,7643 MDL No.: MFCD00011405

Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330 °C
Melting Pt: 681 °C
Density: 3.12
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR